4-(4'-Methoxy-4-biphenylylsulphonylaminomethyl)benzoic acid 96%

Supplier: Thermo Scientific
Warning

H32286.MD H32286.03
ALFAH32286.MDEA 0 SGD
ALFAH32286.MD ALFAH32286.03
4-(4'-Methoxy-4-biphenylylsulphonylaminomethyl)benzoic acid 96%
4-(4'-Methoxy-4-biphenylylsulphonylaminomethyl)benzoic acid

Purity: 96% CAS No.: 885269-44-3 Molecular Formula: C21H19NO5S Formula Weight: 397.45 Melting Point: 250° MDL No.: MFCD06409459

Formula: C₂₁H₁₉NO₅S
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD06409459
CAS Number: 885269-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR