6-Chloro-3-iodo-4H-chromen-4-one 97%
Supplier: Thermo Scientific
H32290.MD
H32290.03
ALFAH32290.MDEA
0
SGD
ALFAH32290.MD
ALFAH32290.03
6-Chloro-3-iodo-4H-chromen-4-one 97%
6-Chloro-3-iodo-4H-chromen-4-one
Purity: 97%
Molecular Formula: C9H4ClIO2
Formula Weight: 306.49
Melting Point: 143-146°
MDL No.: MFCD09475515
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...