4-(2-Chlorophenoxy)benzenesulphonyl chloride 96%

Supplier: Thermo Scientific
Danger

H32346.MD H32346.03 H32346.06
ALFAH32346.MDEA 0 SGD
ALFAH32346.MD ALFAH32346.03 ALFAH32346.06
4-(2-Chlorophenoxy)benzenesulphonyl chloride 96%
4-(2-Chlorophenoxy)benzenesulphonyl chloride

Purity: 96% CAS No.: 610277-84-4 Molecular Formula: C12H8Cl2O3S Formula Weight: 303.17 UN No.: UN3261 MDL No.: MFCD01631899

Formula: C₁₂H₈Cl₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD01631899
CAS Number: 610277-84-4
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR