4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine 96%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-5,6,7,8-tetrahydrobenzo[4,5]thieno[2,3-d]pyrimidine

H32353.03 H32353.06
ALFAH32353.03EA 0 SGD
ALFAH32353.03 ALFAH32353.06
4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine 96%
4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine

Purity: 96% CAS No.: 40493-18-3 Molecular Formula: C10H9ClN2 Formula Weight: 192.65 Melting Point: 95-96° MDL No.: MFCD00463624

Formula: C₁₀H₉ClN₂
MW: 192,65 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00463624
CAS Number: 40493-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR