N-(4'-Methoxy-4-biphenylylsulphonyl)-β-alanine 96%

Supplier: Thermo Scientific
Warning

H32459.MD H32459.03
ALFAH32459.MDEA 0 SGD
ALFAH32459.MD ALFAH32459.03
N-(4'-Methoxy-4-biphenylylsulphonyl)-β-alanine 96%
N-(4'-Methoxy-4-biphenylylsulphonyl)-beta-alanine

Purity: 96% CAS No.: 885269-51-2 Molecular Formula: C16H17NO5S Formula Weight: 335.38 Melting Point: 189-190° MDL No.: MFCD05662691

Formula: C₁₆H₁₇NO₅S
MW: 335,38 g/mol
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD05662691
CAS Number: 885269-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR