1,1'-Diethyl-4,4'-carbocyanine bromide 96%
Supplier: Thermo Scientific
|
Warning
|
H32481.MD
H32481.03
ALFAH32481.MDEA
0
SGD
ALFAH32481.MD
ALFAH32481.03
1,1'-Diethyl-4,4'-carbocyanine bromide 96%
1,1'-Diethyl-4,4'-carbocyanine bromide
Purity: 96%
CAS No.: 19764-88-6
Molecular Formula: C25H25N2Br
Formula Weight: 433.39
MDL No.: MFCD06409816
Formula:
C₂₅H₂₅N₂Br MW: 433,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06409816 CAS Number: 19764-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...