N-(4'-Methoxy-4-biphenylylsulphonyl)-L-phenylalanine 96%
Supplier: Thermo Scientific
|
Warning
|
H32511.MD
H32511.03
ALFAH32511.MDEA
0
SGD
ALFAH32511.MD
ALFAH32511.03
N-(4'-Methoxy-4-biphenylylsulphonyl)-L-phenylalanine 96%
N-(4'-Methoxy-4-biphenylylsulphonyl)-L-phenylalanine
Purity: 96%
Molecular Formula: C22H21NO5S
Formula Weight: 411.48
Melting Point: 178-186°
Optical Rotation: -17° (c=1 in DMF)
MDL No.: MFCD06409467
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...