3-[4-(4-Methoxyphenyl)phenylsulphonamido]benzoic acid

Supplier: Thermo Scientific
Warning

H32583.MD H32583.03
ALFAH32583.MDEA 0 SGD
ALFAH32583.MD ALFAH32583.03
3-[4-(4-Methoxyphenyl)phenylsulphonamido]benzoic acid
3-[4-(4-Methoxyphenyl)phenylsulphonamido]benzoic acid

CAS No.: 885269-91-0 Molecular Formula: C20H17NO5S Formula Weight: 383.43 Melting Point: 235-236° MDL No.: MFCD06409494

Formula: C₂₀H₁₇NO₅S
MW: 383,43 g/mol
Melting Pt: 235…236 °C
Storage Temperature: Ambient
MDL Number: MFCD06409494
CAS Number: 885269-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR