3-Chloro-2,5-difluoropyridine 97%

Supplier: Thermo Scientific
Danger

H32640.MD H32640.03
ALFAH32640.MDEA 0 SGD
ALFAH32640.MD ALFAH32640.03
3-Chloro-2,5-difluoropyridine 97%
3-Chloro-2,5-difluoropyridine

Purity: 97% CAS No.: 851179-00-5 Molecular Formula: C5H2ClF2N Formula Weight: 149.53 Refractive Index: 1.4800 UN No.: UN2924 MDL No.: MFCD09998887

Formula: C₅H₂ClF₂N
MW: 149,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998887
CAS Number: 851179-00-5
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR