1-(4'-Methoxy-4-biphenylylsulphonylamino)cyclohexanecarboxylic acid 96%

Supplier: Thermo Scientific
Warning

H32698.MD H32698.03
ALFAH32698.MDEA 0 SGD
ALFAH32698.MD ALFAH32698.03
1-(4'-Methoxy-4-biphenylylsulphonylamino)cyclohexanecarboxylic acid 96%
1-(4'-Methoxy-4-biphenylylsulphonylamino)cyclohexanecarboxylic acid

Purity: 96% CAS No.: 885269-48-7 Molecular Formula: C20H23NO5S Formula Weight: 389.47 Melting Point: 230° MDL No.: MFCD06409463

Formula: C₂₀H₂₃NO₅S
MW: 389,47 g/mol
Melting Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD06409463
CAS Number: 885269-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR