2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine hydrochloride

H32717.MD H32717.03
ALFAH32717.MDEA 0 SGD
ALFAH32717.MD ALFAH32717.03
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride 95%
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride

Purity: 95% Molecular Formula: C7H7F3N2.HCl Formula Weight: 212.60 MDL No.: MFCD06739079

Formula: C₇H₇F₃N₂·HCl
MW: 212.60 g/mol
Melting Pt: 97…102 °C
Storage Temperature: Ambient
MDL Number: MFCD06739079
CAS Number: 1138011-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR