4-(4'-Methoxy-4-biphenylylsulphonylamino)benzoic acid 96%

Supplier: Thermo Scientific
Warning

H32828.MD H32828.03
ALFAH32828.MDEA 0 SGD
ALFAH32828.MD ALFAH32828.03
4-(4'-Methoxy-4-biphenylylsulphonylamino)benzoic acid 96%
4-(4'-Methoxy-4-biphenylylsulphonylamino)benzoic acid

Purity: 96% CAS No.: 885269-42-1 Molecular Formula: C20H17NO5S Formula Weight: 383.43 Melting Point: 250° MDL No.: MFCD06409458

Formula: C₂₀H₁₇NO₅S
MW: 383,43 g/mol
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD06409458
CAS Number: 885269-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR