(2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one 95%
Supplier: Thermo Scientific
H32875.MD
H32875.03
H32875.06
ALFAH32875.MDEA
0
SGD
ALFAH32875.MD
ALFAH32875.03
ALFAH32875.06
(2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one 95%
(2E,4E)-1-Phenyl-4-(1,3,3-trimethyl-2-indolinylidene)-2-buten-1-one
Purity: 95%
CAS No.: 14575-25-8
Molecular Formula: C21H21NO
Formula Weight: 303.41
Melting Point: 156-162°
MDL No.: MFCD06409497
Formula:
C₂₁H₂₁NO MW: 303,41 g/mol Melting Pt: 156…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD06409497 CAS Number: 14575-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...