4(5)-Cyanomethylimidazole 97%

Supplier: Thermo Scientific
Danger

H32992.03 H32992.06
ALFAH32992.03EA 255 SGD
ALFAH32992.03 ALFAH32992.06
4(5)-Cyanomethylimidazole 97%
4(5)-Cyanomethylimidazole

Purity: 97% CAS No.: 18502-05-1 Molecular Formula: C5H5N3 Formula Weight: 107.11 Odour: Odourless Melting Point: 136-138° UN No.: UN3439 MDL No.: MFCD00070306

Formula: C₅H₅N₃
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00070306
CAS Number: 18502-05-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR