1-Chloromethyl-4-n-octylbenzene 96%

Supplier: Thermo Scientific
Warning

H33122.MD H33122.03
ALFAH33122.MDEA 0 SGD
ALFAH33122.MD ALFAH33122.03
1-Chloromethyl-4-n-octylbenzene 96%
1-Chloromethyl-4-n-octylbenzene

Purity: 96% CAS No.: 40016-26-0 Molecular Formula: C15H23Cl Formula Weight: 238.80 MDL No.: MFCD12923001

Formula: C₁₅H₂₃Cl
MW: 238,80 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923001
CAS Number: 40016-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR