(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%

Supplier: Thermo Fisher Scientific
Warning

H33149.03 H33149.06
ALFAH33149.03EA 0 SGD
ALFAH33149.03 ALFAH33149.06
(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt

Purity: 95% CAS No.: 1268340-94-8 Molecular Formula: C5H11BF3NO Formula Weight: 168.95 Melting Point: 170-172° MDL No.: MFCD20134168

Formula: C₅H₁₁BF₃NO
MW: 168.95 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD20134168
CAS Number: 1268340-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR