3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid 96%

Supplier: Thermo Scientific
Warning

H33253.03 H33253.06
ALFAH33253.03EA 0 SGD
ALFAH33253.03 ALFAH33253.06
3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid 96%
3-(Ethoxycarbonyl)-2-hydroxy-5,6,7,8-tetrahydro-1-naphthoic acid

Purity: 96% CAS No.: 948006-26-6 Molecular Formula: C14H16O5 Formula Weight: 264.28 Melting Point: 172-173° MDL No.: MFCD09258890

Formula: C₁₄H₁₆O₅
MW: 264,28 g/mol
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD09258890
CAS Number: 948006-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR