2-Amino-5-hydroxymethylpyridine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

H33416.MD H33416.03 H33416.06
ALFAH33416.MDEA 0 SGD
ALFAH33416.MD ALFAH33416.03 ALFAH33416.06
2-Amino-5-hydroxymethylpyridine 97%
2-Amino-5-hydroxymethylpyridine

Purity: 97% CAS No.: 113293-71-3 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 113-118° MDL No.: MFCD06659089

Formula: C₆H₈N₂O
MW: 124,14 g/mol
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD06659089
CAS Number: 113293-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR