2-Aminopyridine-6-methanol 97%

Supplier: Thermo Scientific
Warning

Synonyms: (6-Aminopyridin-2-yl)methanol

H33485.03 H33485.06
ALFAH33485.03EA 0 SGD
ALFAH33485.03 ALFAH33485.06
2-Aminopyridine-6-methanol 97%
2-Aminopyridine-6-methanol

Purity: 97% CAS No.: 79651-64-2 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 87-92° MDL No.: MFCD09863097

Formula: C₆H₈N₂O
MW: 124,14 g/mol
Melting Pt: 87…92 °C
Storage Temperature: Ambient
MDL Number: MFCD09863097
CAS Number: 79651-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR