N-(3-Pyridyl)pivalamide 98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,2-Dimethyl-N-pyridin-3-yl-propionamide, 2,2-Dimehtyl-N-pyridin-3-yl-propionamide

H33510.03 H33510.06
ALFAH33510.03EA 0 SGD
ALFAH33510.03 ALFAH33510.06
N-(3-Pyridyl)pivalamide 98%
N-(3-Pyridyl)pivalamide

Purity: 98% CAS No.: 70298-88-3 Molecular Formula: C10H14N2O Formula Weight: 178.23 Melting Point: 129-131° MDL No.: MFCD00996243

Formula: C₁₀H₁₄N₂O
MW: 178,23 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00996243
CAS Number: 70298-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR