(5-Bromofuro[2,3-b]pyridin-2-yl)methanol ≥98%
Supplier: Thermo Scientific
|
Warning
|
H33622.03
H33622.06
ALFAH33622.03EA
0
SGD
ALFAH33622.03
ALFAH33622.06
(5-Bromofuro[2,3-b]pyridin-2-yl)methanol ≥98%
(5-Bromofuro[2,3-b]pyridin-2-yl)methanol
Purity: 98%
CAS No.: 1188990-02-4
Molecular Formula: C8H6BrNO2
Formula Weight: 228.05
Melting Point: 133-135°
MDL No.: MFCD13176658
Formula:
C₈H₆BrNO₂ Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD13176658 CAS Number: 1188990-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...