2-(Trifluoromethyl)quinoline 97%
Supplier: Thermo Scientific
|
Warning
|
H33740.MD
H33740.03
ALFAH33740.MDEA
0
SGD
ALFAH33740.MD
ALFAH33740.03
2-(Trifluoromethyl)quinoline 97%
2-(Trifluoromethyl)quinoline
Purity: 97%
CAS No.: 347-42-2
Molecular Formula: C10H6F3N
Formula Weight: 197.16
Melting Point: 59-62°
MDL No.: MFCD01862002
Formula:
C₁₀H₆F₃N Melting Pt: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862002 CAS Number: 347-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...