4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine 96%
Supplier: Thermo Scientific
|
Warning
|
H33770.03
H33770.06
ALFAH33770.03EA
234
SGD
ALFAH33770.03
ALFAH33770.06
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine 96%
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Purity: 96%
CAS No.: 108831-68-1
Molecular Formula: C8H7ClN2S
Formula Weight: 198.68
Melting Point: 109-110°
MDL No.: MFCD00464832
Formula:
C₈H₇ClN₂S Melting Pt: 109…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464832 CAS Number: 108831-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...