2,3,5,6-Tetrafluoropyridine-4-acetic acid 98%

Supplier: Thermo Scientific
Warning

H33785.MD H33785.03
ALFAH33785.MDEA 0 SGD
ALFAH33785.MD ALFAH33785.03
2,3,5,6-Tetrafluoropyridine-4-acetic acid 98%
2,3,5,6-Tetrafluoropyridine-4-acetic acid

Purity: 98% CAS No.: 65697-73-6 Molecular Formula: C7H3F4NO2 Formula Weight: 209.10 Melting Point: 145-149° MDL No.: MFCD02944320

Formula: C₇H₃F₄NO₂
MW: 209,10 g/mol
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD02944320
CAS Number: 65697-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR