3,5-Dichloro-2,6-difluoro-4-pyridineacetic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H33892.MD
H33892.03
ALFAH33892.MDEA
0
SGD
ALFAH33892.MD
ALFAH33892.03
3,5-Dichloro-2,6-difluoro-4-pyridineacetic acid 97%
3,5-Dichloro-2,6-difluoro-4-pyridineacetic acid
Purity: 97%
Molecular Formula: C7H3Cl2F2NO2
Formula Weight: 242.01
Melting Point: 153-157°
MDL No.: MFCD20040118
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...