4-[4-(2-Chlorophenoxy)phenylsulphonylamino]benzoic acid 96%
Supplier: Thermo Scientific
|
Warning
|
H34000.MD
H34000.03
ALFAH34000.MDEA
0
SGD
ALFAH34000.MD
ALFAH34000.03
4-[4-(2-Chlorophenoxy)phenylsulphonylamino]benzoic acid 96%
4-[4-(2-Chlorophenoxy)phenylsulphonylamino]benzoic acid
Purity: 96%
CAS No.: 606944-48-3
Molecular Formula: C19H14ClNO5S
Formula Weight: 403.84
Melting Point: 224-226°
MDL No.: MFCD06261211
Formula:
C₁₉H₁₄ClNO₅S MW: 403,84 g/mol Melting Pt: 224…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD06261211 CAS Number: 606944-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...