tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-BOC protected
H34134.03
H34134.06
ALFAH34134.03EA
0
SGD
ALFAH34134.03
ALFAH34134.06
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Purity: 97%
CAS No.: 147804-30-6
Molecular Formula: C11H19NO3
Formula Weight: 213.28
Melting Point: 56-58°
MDL No.: MFCD07779385
Formula:
C₁₁H₁₉NO₃ MW: 213,28 g/mol Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD07779385 CAS Number: 147804-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...