3-Bromo-4-iodopyridine 97%

Supplier: Thermo Scientific
Warning

H34203.03 H34203.MD
ALFAH34203.03EA 0 SGD
ALFAH34203.03 ALFAH34203.MD
3-Bromo-4-iodopyridine 97%
3-Bromo-4-iodopyridine

Purity: 97% CAS No.: 89167-19-1 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Melting Point: 111-115° MDL No.: MFCD11110263

Formula: C₅H₃BrIN
MW: 283,89 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD11110263
CAS Number: 89167-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR