3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
DiSC2(3), 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzothiazolylidene]-1-propenyl]benzothiazolium Iodide, 3,3-Diethylthiacarbocyanine iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]thiazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]thiazol-3-ium iodide
Purity: 96%
CAS No.: 905-97-5
Molecular Formula: C21H21IN2S2
Formula Weight: 492.45
Melting Point: ca 250° dec
MDL No.: MFCD00011956
Formula:
C₂₁H₂₁IN₂S₂ MW: 492,44 g/mol Melting Pt: ∼ 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011956 CAS Number: 905-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...