2-Hydrazino-4-methylquinoline 96%

Supplier: Thermo Scientific
Warning

H34273.03 H34273.MD
ALFAH34273.03EA 0 SGD
ALFAH34273.03 ALFAH34273.MD
2-Hydrazino-4-methylquinoline 96%
2-Hydrazino-4-methylquinoline

Purity: 96% CAS No.: 21703-52-6 Molecular Formula: C10H11N3 Formula Weight: 173.22 MDL No.: MFCD00818533

Formula: C₁₀H₁₁N₃
MW: 173,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00818533
CAS Number: 21703-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR