3-(4-Cyanophenyl)propanoic acid 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-(4-Cyanophenyl)propionic acid
H34297.03
H34297.06
ALFAH34297.03EA
0
SGD
ALFAH34297.03
ALFAH34297.06
3-(4-Cyanophenyl)propanoic acid 96%
3-(4-Cyanophenyl)propanoic acid
Purity: 96%
CAS No.: 42287-94-5
Molecular Formula: C10H9NO2
Formula Weight: 175.19
UN No.: UN3439
MDL No.: MFCD08234823
Formula:
C₁₀H₉NO₂ MW: 175,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08234823 CAS Number: 42287-94-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...