Julolidine hydrobromide 97%
Supplier: Thermo Scientific
|
Danger
|
H36298.14
H36298.22
ALFAH36298.14EA
0
SGD
ALFAH36298.14
ALFAH36298.22
Julolidine hydrobromide 97%
Julolidine hydrobromide
Purity: 97%
CAS No.: 83646-41-7
Molecular Formula: C12H16BrN
Formula Weight: 254.17
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Pale pink
Melting Point: 239-242°
UN No.: UN2923
MDL No.: MFCD00198009
Formula:
C₁₂H₁₆BrN MW: 254,171 g/mol Melting Pt: 239…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00198009 CAS Number: 83646-41-7 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...