2,2-Dimethyl-N-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)pyridin-2-yl]propionamide 98%

Supplier: Thermo Fisher Scientific
Warning

H50015.03 H50015.MD
ALFAH50015.03EA 0 SGD
ALFAH50015.03 ALFAH50015.MD
2,2-Dimethyl-N-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)pyridin-2-yl]propionamide 98%
2,2-Dimethyl-N-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)pyridin-2-yl]propionamide

Purity: 98% CAS No.: 532391-30-3 Molecular Formula: C16H25BN2O3 Formula Weight: 304.20 Melting Point: 155° MDL No.: MFCD06658997

Formula: C₁₆H₂₅BN₂O₃
MW: 304.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658997
CAS Number: 532391-30-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR