N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide 98%

Supplier: Thermo Scientific
Warning

H50035.03 H50035.MD
ALFAH50035.03EA 448 SGD
ALFAH50035.03 ALFAH50035.MD
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide 98%
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide

Purity: 98% CAS No.: 169205-93-0 Molecular Formula: C10H14N2O2 Formula Weight: 194.24 MDL No.: MFCD07776841

Formula: C₁₀H₁₄N₂O₂
MW: 194,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07776841
CAS Number: 169205-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR