5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Bromo-7-azaindole
H50040.03
H50040.MD
ALFAH50040.03EA
120
SGD
ALFAH50040.03
ALFAH50040.MD
5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Purity: 96%
CAS No.: 183208-35-7
Molecular Formula: C7H5BrN2
Formula Weight: 197.04
Melting Point: 178-179°
MDL No.: MFCD06659677
Formula:
C₇H₅BrN₂ MW: 197,04 g/mol Melting Pt: 178…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659677 CAS Number: 183208-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...