2-(Boc-amino)pyridine-5-carboxaldehyde 97%

Supplier: Thermo Scientific
Warning

Synonyms: tert-Butyl (5-formylpyridin-2-yl)carbamate, 6-Aminonicotinaldehyde, 6-BOC protected

H50075.03 H50075.MD
ALFAH50075.03EA 581 SGD
ALFAH50075.03 ALFAH50075.MD
2-(Boc-amino)pyridine-5-carboxaldehyde 97%
2-(Boc-amino)pyridine-5-carboxaldehyde

Purity: 97% CAS No.: 199296-40-7 Molecular Formula: C11H14N2O3 Formula Weight: 222.25 MDL No.: MFCD08064228

Formula: C₁₁H₁₄N₂O₃
MW: 222,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064228
CAS Number: 199296-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR