1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid 96%
Supplier: Thermo Scientific
|
Warning
|
H50086.03
H50086.MD
ALFAH50086.03EA
545
SGD
ALFAH50086.03
ALFAH50086.MD
1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid 96%
1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid
Purity: 96%
CAS No.: 1210863-93-6
Molecular Formula: C12H19NO6
Formula Weight: 273.29
MDL No.: MFCD07781134
Formula:
C₁₂H₁₉NO₆ MW: 273,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781134 CAS Number: 1210863-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...