4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester 99%

Supplier: Thermo Scientific
Warning

Synonyms: tert-Butyl 4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine-1-carboxylate, 6-[4-(tert-Butoxycarbonyl)piperazin-1-yl]pyridine-3-boronic acid, pinacol ester

H50145.03 H50145.MD
ALFAH50145.03EA 374 SGD
ALFAH50145.03 ALFAH50145.MD
4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester 99%
4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester

Purity: 99% CAS No.: 496786-98-2 Molecular Formula: C20H32BN3O4 Formula Weight: 389.31 MDL No.: MFCD04039875

Formula: C₂₀H₃₂BN₃O₄
MW: 389,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039875
CAS Number: 496786-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR