Ethyl-2-chloro-3-pyridylglyoxylate 97%

Supplier: Thermo Scientific

H50153.03 H50153.06
ALFAH50153.03EA 402 SGD
ALFAH50153.03 ALFAH50153.06
Ethyl-2-chloro-3-pyridylglyoxylate 97%
Ethyl-2-chloro-3-pyridylglyoxylate

Purity: 97% CAS No.: 902837-56-3 Molecular Formula: C9H8ClNO3 Formula Weight: 213.62 MDL No.: MFCD07781194

Formula: C₉H₈ClNO₃
MW: 213,617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781194
CAS Number: 902837-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR