4,4,4-Trichlorobutyric acid

Supplier: Thermo Scientific
Danger

H50219.03 H50219.06
ALFAH50219.03EA 214 SGD
ALFAH50219.03 ALFAH50219.06
4,4,4-Trichlorobutyric acid
4,4,4-Trichlorobutyric acid

CAS No.: 2345-32-6 Molecular Formula: Cl3CCH2CH2CO2H Formula Weight: 191.44 UN No.: UN3261 MDL No.: MFCD00100736

Formula: Cl₃CCH₂CH₂CO₂H
MW: 191,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100736
CAS Number: 2345-32-6
EINECS: 219-065-3
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR