3-(Methylaminomethyl)pyridine 98%

Supplier: Thermo Scientific
Warning

Synonyms: N-Methyl-1-(pyridin-3-yl)methanamine, N-Methyl-3-picolylamine, N-(3-Pyridylmethyl)methylamine

H50262.03 H50262.06
ALFAH50262.03EA 105 SGD
ALFAH50262.03 ALFAH50262.06
3-(Methylaminomethyl)pyridine 98%
3-(Methylaminomethyl)pyridine

Purity: 98% CAS No.: 20173-04-0 Molecular Formula: C7H10N2 Formula Weight: 122.17 Boiling Point: 93-95°/9mm MDL No.: MFCD00023610

Formula: C₇H₁₀N₂
MW: 122,17 g/mol
Boiling Pt: 93…95 °C (9 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00023610
CAS Number: 20173-04-0
EINECS: 243-561-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR