(4'-Fluoro[1,1'-biphenyl]-2-yl)methanamine 97%

Supplier: Thermo Scientific
Warning

H50297.03 H50297.MD
ALFAH50297.03EA 568 SGD
ALFAH50297.03 ALFAH50297.MD
(4'-Fluoro[1,1'-biphenyl]-2-yl)methanamine 97%
(4'-Fluoro[1,1'-biphenyl]-2-yl)methanamine

Purity: 97% CAS No.: 884504-18-1 Molecular Formula: C13H12FN Formula Weight: 201.25 MDL No.: MFCD05864430

Formula: C₁₃H₁₂FN
Storage Temperature: Ambient
MDL Number: MFCD05864430
CAS Number: 884504-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR