4'-Fluoro-3-biphenylmethylamine hydrochloride 99%

Supplier: Thermo Scientific
Warning

Synonyms: (4'-Fluoro-[1,1'-biphenyl]-3-yl)methanamine hydrochloride

H50298.03 H50298.MD
ALFAH50298.03EA 561 SGD
ALFAH50298.03 ALFAH50298.MD
4'-Fluoro-3-biphenylmethylamine hydrochloride 99%
4'-Fluoro-3-biphenylmethylamine hydrochloride

Purity: 99% CAS No.: 1195901-44-0 Molecular Formula: C13H12FN.HCl Formula Weight: 237.71 MDL No.: MFCD02089423

Formula: C₁₃H₁₂FN·HCl
MW: 237,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02089423
CAS Number: 1195901-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR