4'-Methylbiphenyl-4-sulphonyl chloride 98%

Supplier: Thermo Scientific
Danger

Synonyms: 4'-Methyl-[1,1'-biphenyl]-4-sulphonyl chloride

H50302.03 H50302.MD
ALFAH50302.03EA 0 SGD
ALFAH50302.03 ALFAH50302.MD
4'-Methylbiphenyl-4-sulphonyl chloride 98%
4'-Methylbiphenyl-4-sulphonyl chloride

Purity: 98% CAS No.: 101366-51-2 Molecular Formula: C13H11ClO2S Formula Weight: 266.74 UN No.: UN3261 MDL No.: MFCD01631922

Formula: C₁₃H₁₁ClO₂S
MW: 266,74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631922
CAS Number: 101366-51-2
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR