5-Cyclopropyl-1H-pyrazole-3-carboxylic acid 94%
Supplier: Thermo Scientific
|
Warning
|
H50306.03
H50306.MD
ALFAH50306.03EA
480
SGD
ALFAH50306.03
ALFAH50306.MD
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid 94%
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Purity: 94%
CAS No.: 401629-04-7
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: ca 175° dec.
MDL No.: MFCD04967194
Formula:
C₇H₈N₂O₂ MW: 152,15 g/mol Melting Pt: ∼ 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04967194 CAS Number: 890591-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...