Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H50318.03
H50318.MD
ALFAH50318.03EA
380
SGD
ALFAH50318.03
ALFAH50318.MD
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Purity: 97%
CAS No.: 25940-35-6
Molecular Formula: C7H5N3O2
Formula Weight: 163.14
Melting Point: ca 180° dec.
MDL No.: MFCD01975194
Formula:
C₇H₅N₃O₂ Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01975194 CAS Number: 25940-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...