2-Methyl-1,3-benzothiazol-6-amine 99%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Amino-2-methyl-1,3-benzothiazole

H50319.03 H50319.MD
ALFAH50319.03EA 296 SGD
ALFAH50319.03 ALFAH50319.MD
2-Methyl-1,3-benzothiazol-6-amine 99%
2-Methyl-1,3-benzothiazol-6-amine

Purity: 99% CAS No.: 2941-62-0 Molecular Formula: C8H8N2S Formula Weight: 164.22 MDL No.: MFCD00160076

Formula: C₈H₈N₂S
MW: 164,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160076
CAS Number: 2941-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR