4-Methoxy-3-methylbenzyl alcohol

Supplier: Thermo Scientific
Warning

H50344.03 H50344.MD
ALFAH50344.03EA 567 SGD
ALFAH50344.03 ALFAH50344.MD
4-Methoxy-3-methylbenzyl alcohol
4-Methoxy-3-methylbenzyl alcohol

CAS No.: 114787-91-6 Molecular Formula: C9H12O2 Formula Weight: 152.19 MDL No.: MFCD00060357

Formula: C₉H₁₂O₂
MW: 152,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060357
CAS Number: 114787-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR