5-Chlorosulphonyl-2-hydroxybenzoic acid 97%

Supplier: Thermo Scientific
Danger

H50374.03 H50374.06
ALFAH50374.03EA 210 SGD
ALFAH50374.03 ALFAH50374.06
5-Chlorosulphonyl-2-hydroxybenzoic acid 97%
5-Chlorosulphonyl-2-hydroxybenzoic acid

Purity: 97% CAS No.: 17243-13-9 Molecular Formula: C7H5ClO5S Formula Weight: 236.63 Melting Point: 171-174° UN No.: UN2923 MDL No.: MFCD00052974

Formula: C₇H₅ClO₅S
MW: 236,63 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00052974
CAS Number: 17243-13-9
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR