3-Fluoro-4-methoxybenzenesulphonyl chloride 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Fluoro-4-methoxybenzene-1-sulphonyl chloride
H50383.03
H50383.06
ALFAH50383.03EA
213
SGD
ALFAH50383.03
ALFAH50383.06
3-Fluoro-4-methoxybenzenesulphonyl chloride 98%
3-Fluoro-4-methoxybenzenesulphonyl chloride
Purity: 98%
CAS No.: 67475-55-2
Molecular Formula: C7H6ClFO3S
Formula Weight: 224.64
Melting Point: 77-80°
UN No.: UN3261
MDL No.: MFCD02091380
Formula:
C₇H₆ClFO₃S MW: 224,64 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD02091380 CAS Number: 67475-55-2 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...